Learn More
Nova Biomedical Corporation Nova Biomedical FAST CDV Reagent Pack, Supports Trypan Blue Exclusion, RFID Technology, 1 Pack
Catalog No. NC3592589
Encompass
The Nova Biomedical FAST CDV Reagent Pack enhances laboratory workflows by providing a high-capacity solution for the BioProfile FAST CDV analyzer, facilitating rapid and accurate cell density and viability measurements. Featuring RFID technology for auto-recognition and an integrated waste bag, this reagent pack minimizes manual handling and maintenance, ensuring efficient analysis with over 45 samples per hour. It's designed for reliability and ease of use, setting it apart from other reagents.
Catalog No. NC3592589 Supplier Nova Biomedical Corporation Supplier No. 64739
The Nova Biomedical FAST CDV Reagent Pack enhances laboratory workflows by providing a high-capacity solution for the BioProfile FAST CDV analyzer, facilitating rapid and accurate cell density and viability measurements. Featuring RFID technology for auto-recognition and an integrated waste bag, this reagent pack minimizes manual handling and maintenance, ensuring efficient analysis with over 45 samples per hour. It's designed for reliability and ease of use, setting it apart from other reagents.