Learn More
Meinhard Elemental Scientific FAST connector for F5 (original) Meinhard Nebulizer, 0.25mm i.d. (green marker),attaches to port #3 of the FAST valve, 30cm long
Catalog No. NC2736401
Encompass
FAST connector for F5 (original) Meinhard Nebulizer, 0.25mm i.d. (green marker),attaches to port #3 of the FAST valve, 30cm long
Catalog No. NC2736401 Supplier Meinhard Elemental Scientific Supplier No. FF52530
FAST connector for F5 (original) Meinhard Nebulizer, 0.25mm i.d. (green marker),attaches to port #3 of the FAST valve, 30cm long