Learn More
Meinhard Elemental Scientific AST connector for MEINHARD® Plus Series Nebulizer F2 (0.25mm i.d., greenmarker) Attaches to port #3 of the FAST valve, 30cm long
Catalog No. NC2736398
Encompass
FAST connector for MEINHARD® Plus Series Nebulizer F2 (0.25mm i.d., greenmarker) Attaches to port #3 of the FAST valve, 30cm long
Catalog No. NC2736398 Supplier Meinhard Elemental Scientific Supplier No. FF22530
FAST connector for MEINHARD® Plus Series Nebulizer F2 (0.25mm i.d., greenmarker) Attaches to port #3 of the FAST valve, 30cm long