Learn More
Grainger Fast Fill Funnel 2qt. 1-1/16 Dia Spout
Catalog No. 502750465
Encompass_Preferred
The Funnel King Fast Fill Spout Funnel is a polyethylene funnel designed for fast pouring. It has a 6 inch diameter top opening that tapers down to a 1-1/4 inch diameter spout. The overall height is 9 inches. This durable, chemical resistant funnel has a ribbed exterior for a non-slip grip. It can transfer up to 5 gallons per minute. The spout has a cap for dust protection. This funnel is ideal for automotive fluids, chemicals, granular materials and powders. It's made in the USA.
Catalog No. 50-275-0465 Supplier Grainger Supplier No. 14N904
The Funnel King Fast Fill Spout Funnel is a polyethylene funnel designed for fast pouring. It has a 6 inch diameter top opening that tapers down to a 1-1/4 inch diameter spout. The overall height is 9 inches. This durable, chemical resistant funnel has a ribbed exterior for a non-slip grip. It can transfer up to 5 gallons per minute. The spout has a cap for dust protection. This funnel is ideal for automotive fluids, chemicals, granular materials and powders. It's made in the USA.