Learn More
Epredia™ Fast Flex™ Holding Solution
Catalog No. 23859100 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5L
2.5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-859-100 2.5 gal.
98-001 5L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-859-100 Supplier Epredia™ Supplier No. 9800
Add to Cart
Add to Cart
Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
Specifications
Quantity 2.5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov