Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Fast Flex™ Holding Solution

Catalog No. 98001 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5L
2.5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-001 5L
23-859-100 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 98-001 Supplier Epredia™ Supplier No. 98001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.

  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives

Specifications

Product Type Holding Solution
Quantity 5L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.