Learn More
Epredia™ Fast-Flex™ Processing Reagents
Catalog No. 23859101 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Xylene-based Clearant
Catalog No. Product Type
23-859-101 Xylene-based Clearant
1 options
Catalog No. 23-859-101 Supplier Epredia™ Supplier No. 9801
Add to Cart
Add to Cart
Improve the quality of processed tissue with these processing reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
  • Reduces Tissue processing time by 80%
  • Decreases the amount of reagents used up to 75%
  • Simplifies reagent management by eliminating the need to rotate reagents
  • Compatible with H&E, IHC, Special Stains and In situ
Includes:
One 5L bottle each: Fixative (Solution 1), Dehydrant (Solution 2) and Clearing Reagent (Solution 3).
Specifications
Product Type Xylene-based Clearant
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov