Learn More
Waters Corp Waters Column, 50 Å, 7 μm, 7.8 mm X 150 mm
Catalog No. 50784977 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-performance, reversed-phase column is designed for the separation of glycerol, organic acids, ethanol, and small ions from fruit juices, delivering exceptional separation efficiency.
  • Separates glycerol, organic acids, ethanol, and small ions from fruit juices.
  • Silica-based material with enhanced hydrophobicity.
  • Pore size of 50 Å and particle size of 7 μm.
  • Measures 7.8 mm × 150 mm; one column per package.
  • Ideal for LC, bioanalysis, and pharmaceutical applications.
  • Provides excellent compound separation, high resolution, and reproducible data.
  • Compatible with various mobile phases for flexible method development.
  • Ensures consistent and stable performance with evenly sized spherical particles.
Catalog No. 50-784-977 Supplier Waters Corp Supplier No. WAT010639
This high-performance, reversed-phase column is designed for the separation of glycerol, organic acids, ethanol, and small ions from fruit juices, delivering exceptional separation efficiency.
  • Separates glycerol, organic acids, ethanol, and small ions from fruit juices.
  • Silica-based material with enhanced hydrophobicity.
  • Pore size of 50 Å and particle size of 7 μm.
  • Measures 7.8 mm × 150 mm; one column per package.
  • Ideal for LC, bioanalysis, and pharmaceutical applications.
  • Provides excellent compound separation, high resolution, and reproducible data.
  • Compatible with various mobile phases for flexible method development.
  • Ensures consistent and stable performance with evenly sized spherical particles.