Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast garnet GBC base, MP Biomedicals™

Catalog No. ICN10169080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10169080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10169080 Supplier MP Biomedicals, Inc Supplier No. 0210169080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azo dye used in various laboratory and industrial applications.

  • Chemical Name: Fast garnet GBC base
  • Molecular Formula: C14H15N3
  • Formula Weight: 225.3
  • Physical Form: Powder
  • CAS Registry Number: 97-56-3
  • IUPAC Name: 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
  • Synonym: Oil yellow 170
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym Oil yellow 170, Solvent yellow 3
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C

Specifications

Color Orange
pH 5 to 7
Quantity 100 g
Physical Form Powder
Formula Weight 225.3
Chemical Name or Material Fast garnet GBC base
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.