Learn More
Medchemexpress LLC Fast garnet GBC sulfate salt | 101-89-3 | MFCD00011883 | 90.0% | 334.35 g·mol⁻¹ | C14H14N4O4S | 100 MG
SDP
Catalog No. 5000281874
Encompass_Preferred
Fast garnet GBC sulfate salt, 90% is a chromogenic substrate for alkaline phosphatase used in histology and biochemical assays. It yields a red-violet azo precipitate at enzyme activity sites and is supplied in small pack sizes for research use, including 50 mg, 100 mg, and 250 mg. CAS 101-89-3; molecular weight 334.35 g·mol⁻¹.
  • Chromogenic substrate for alkaline phosphatase staining and detection.
  • Approximately 90% purity suitable for research applications.
  • Produces a red-violet azo precipitate for clear visualization.
  • Available in small pack sizes for laboratory use.
  • Supplied as the sulfate salt with CAS number 101-89-3.
Catalog No. 50-002-81874 Supplier Medchemexpress LLC Supplier No. HYW328317100MG
Fast garnet GBC sulfate salt, 90% is a chromogenic substrate for alkaline phosphatase used in histology and biochemical assays. It yields a red-violet azo precipitate at enzyme activity sites and is supplied in small pack sizes for research use, including 50 mg, 100 mg, and 250 mg. CAS 101-89-3; molecular weight 334.35 g·mol⁻¹.
  • Chromogenic substrate for alkaline phosphatase staining and detection.
  • Approximately 90% purity suitable for research applications.
  • Produces a red-violet azo precipitate for clear visualization.
  • Available in small pack sizes for laboratory use.
  • Supplied as the sulfate salt with CAS number 101-89-3.