Learn More
Medchemexpress LLC Fast garnet GBC sulfate salt | 101-89-3 | MFCD00011883 | 90.0% | 334.35 g/mol | C14H14N4O4S | 250 MG
SDP
Catalog No. 5000282060
Encompass_Preferred
Fast garnet GBC sulfate salt, 90% is a chromogenic substrate for alkaline phosphatase used to detect and visualize enzyme activity in histochemical and biochemical assays. It is supplied as a brown to black solid powder with a stated purity of 90% and is offered in multiple laboratory-scale pack sizes.
  • Chromogenic substrate for alkaline phosphatase.
  • Provides visible staining to localize enzymatic activity in tissue and cell samples.
  • Supplied as a solid powder, brown to black in appearance.
  • Stated purity of 90.0% supports consistent staining performance.
  • Available in multiple laboratory pack sizes for flexible use.
  • Molecular formula C14H14N4O4S, molecular weight 334.35 g/mol.
Catalog No. 50-002-82060 Supplier Medchemexpress LLC Supplier No. HYW328317250MG
Fast garnet GBC sulfate salt, 90% is a chromogenic substrate for alkaline phosphatase used to detect and visualize enzyme activity in histochemical and biochemical assays. It is supplied as a brown to black solid powder with a stated purity of 90% and is offered in multiple laboratory-scale pack sizes.
  • Chromogenic substrate for alkaline phosphatase.
  • Provides visible staining to localize enzymatic activity in tissue and cell samples.
  • Supplied as a solid powder, brown to black in appearance.
  • Stated purity of 90.0% supports consistent staining performance.
  • Available in multiple laboratory pack sizes for flexible use.
  • Molecular formula C14H14N4O4S, molecular weight 334.35 g/mol.