Learn More
Medchemexpress LLC Fast garnet GBC sulfate salt | 101-89-3 | MFCD00011883 | 90.0% | 334.35 g/mol | C14H14N4O4S | 50 MG
SDP
Catalog No. 5000280795
Encompass_Preferred
Fast garnet GBC sulfate salt, 90% is an azo diazonium dye used as a chromogenic substrate for alkaline phosphatase in histochemical staining and related enzymatic assays. It is supplied as a sulfate salt with high dye content suitable for laboratory staining protocols.
  • Chromogenic substrate for alkaline phosphatase staining
  • High dye content, formulated at 90.0% purity
  • Suitable for histochemistry and enzyme activity assays
  • Available in small laboratory-scale quantities
  • Solid form with molecular formula C14H14N4O4S
Catalog No. 50-002-80795 Supplier Medchemexpress LLC Supplier No. HYW32831750MG
Fast garnet GBC sulfate salt, 90% is an azo diazonium dye used as a chromogenic substrate for alkaline phosphatase in histochemical staining and related enzymatic assays. It is supplied as a sulfate salt with high dye content suitable for laboratory staining protocols.
  • Chromogenic substrate for alkaline phosphatase staining
  • High dye content, formulated at 90.0% purity
  • Suitable for histochemistry and enzyme activity assays
  • Available in small laboratory-scale quantities
  • Solid form with molecular formula C14H14N4O4S