Learn More
Roche Diagnostics KAPA Mouse Genotyping Kits - Non-HotStart PCR, without extraction, 500 x 25 uL
Catalog No. 501965216 Shop All Roche Diagnostics Products
Encompass
KAPA Mouse Gentoyping Kits include KAPA Express Extract Enzyme (1 U/uL), KAPA Express Extract Buffer (10X), and KAPA2G Fast (or Fast HotStart) Genotyping Mix (2X), which contains DNA Polymerase, reaction buffer with loading dye, dNTPs, and MgCl2 (at a final concentration of 1.5 mM). ReadyMixes for PCR are also sold separately. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5216 Supplier Roche Diagnostics Supplier No. 07960921001
KAPA Mouse Gentoyping Kits include KAPA Express Extract Enzyme (1 U/uL), KAPA Express Extract Buffer (10X), and KAPA2G Fast (or Fast HotStart) Genotyping Mix (2X), which contains DNA Polymerase, reaction buffer with loading dye, dNTPs, and MgCl2 (at a final concentration of 1.5 mM). ReadyMixes for PCR are also sold separately. *This item is non returnable.