Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Green Dye Indicator, Reagents

Catalog No. 01335261
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-261 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-261 Supplier Reagents Holdings Llc Supplier No. CF128000500A
Add to Cart
Add to Cart

Fast Green Dye Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fast Green Dye Indicator
CAS 7732-18-5, 56-81-5, 2353-45-9
Packaging Poly Bottle
Specific Gravity 1.0555
Synonym Food Green 3, FD and C Green 3
Color Dark Green
Quantity 500 mL
Physical Form Liquid
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.