Learn More
Cayman Chemical Fast Green FCF 100g
SDP
Catalog No. 502953946
Encompass_Preferred
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)
Catalog No. 50-295-3946 Supplier Cayman Chemical Supplier No. 14335100 G
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)