Learn More
Cayman Chemical Fast Green FCF 25g
SDP
Catalog No. 502954024
Encompass_Preferred
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)
Catalog No. 50-295-4024 Supplier Cayman Chemical Supplier No. 1433525 G
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)