Learn More
Cayman Chemical Fast Green FCF, CAS: 2353-45-9, 5 g
SDP
Catalog No. 502954586
Encompass_Preferred
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)
Catalog No. 50-295-4586 Supplier Cayman Chemical Supplier No. 143355 G
A dye used to stain proteins for IEF and SDS-PAGE; fluoresces near infrared (absorption maximum: 624 nm)