Learn More
Electron Microscopy Sciences Fast Green FCF Solution 0.01% 500 ML
SDP
Catalog No. 5031920
Encompass_Preferred
Used in Lillie's Method for Nucleic Acids (1965) for nuclear chromatin, and chromatin of plasmodia, etc. See references: Lillie, R.D., Histopathologic Technic and Practical Histochemistry, 3rd ed, McGraw-Hill, NY, pp 149-150, c. 1965 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd ed. McGraw-Hill, NY, p. 131, c. 1968.
Catalog No. 50-319-20 Supplier Electron Microscopy Sciences Supplier No. 2677404
Used in Lillie's Method for Nucleic Acids (1965) for nuclear chromatin, and chromatin of plasmodia, etc. See references: Lillie, R.D., Histopathologic Technic and Practical Histochemistry, 3rd ed, McGraw-Hill, NY, pp 149-150, c. 1965 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd ed. McGraw-Hill, NY, p. 131, c. 1968.