Learn More
Electron Microscopy Sciences Fast Green FCF Solution 2.5% in Acetic Acid 500 ML
SDP
Catalog No. 5031958
Encompass_Preferred
Fast Green FCF solution is a ready-to-use, high quality staining solution that saves time and effort and provides consistently uniform results Concentration of2.5% aqeous in 2% acetic acid.
Catalog No. 50-319-58 Supplier Electron Microscopy Sciences Supplier No. 2605325
Fast Green FCF solution is a ready-to-use, high quality staining solution that saves time and effort and provides consistently uniform results Concentration of2.5% aqeous in 2% acetic acid.