Learn More
ABclonal Technology Fast Myosin Skeletal HC
SDP
Supplier:  ABclonal Technology A27146100UL
Encompass_Preferred
Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains Myosin heavy chains are encoded by a multigene family In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated smooth and nonmuscle cells These isoforms show expression that is spatially and temporally regulated during development
Catalog No. 50-000-12505