Learn More
Meridian Life Science Fast qPCR Buffer 4x, 2500UL
SDP
Catalog No. 5000856552
Encompass_Preferred
Fast qPCR Buffer, 4x is a novel buffer system that has been designed to be combined with a high-quality hot-start Taq DNA polymerase such as MDX008, to deliver highly reproducible, accurate assay results under fast thermal cycling conditions, delivering excellent results in fast qPCR assays.
Catalog No. 50-008-56552 Supplier Meridian Life Science Supplier No. MDX0332.5
Fast qPCR Buffer, 4x is a novel buffer system that has been designed to be combined with a high-quality hot-start Taq DNA polymerase such as MDX008, to deliver highly reproducible, accurate assay results under fast thermal cycling conditions, delivering excellent results in fast qPCR assays.