Learn More
Meridian Life Science Fast qPCR Buffer 4x, 50ML
SDP
Catalog No. 5000856314
Encompass_Preferred
Fast qPCR Buffer, 4x is a novel buffer system that has been designed to be combined with a high-quality hot-start Taq DNA polymerase such as MDX008, to deliver highly reproducible, accurate assay results under fast thermal cycling conditions, delivering excellent results in fast qPCR assays.
Catalog No. 50-008-56314 Supplier Meridian Life Science Supplier No. MDX03350
Fast qPCR Buffer, 4x is a novel buffer system that has been designed to be combined with a high-quality hot-start Taq DNA polymerase such as MDX008, to deliver highly reproducible, accurate assay results under fast thermal cycling conditions, delivering excellent results in fast qPCR assays.