Learn More
Meridian Life Science Fast qPCR Mix 5x, 40ML
SDP
Catalog No. 5000856288
Encompass_Preferred
Fast qPCR Mix, 5x has been designed with the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with a fast hot-start polymerase, dNTPs and MgCl2, for highly reproducible, accurate, sensitive qPCR efficiency, even from low abundance targets and scarce samples. With high processivity and robustness, it delivers excellent results in both singleplex and multiplex qPCR assays and confers superior assay performance under fast thermal cycling conditions, allowing more samples to be run in a day with the highest confidence, ideal for high‑throughput assays.
Catalog No. 50-008-56288 Supplier Meridian Life Science Supplier No. MDX07240
Fast qPCR Mix, 5x has been designed with the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with a fast hot-start polymerase, dNTPs and MgCl2, for highly reproducible, accurate, sensitive qPCR efficiency, even from low abundance targets and scarce samples. With high processivity and robustness, it delivers excellent results in both singleplex and multiplex qPCR assays and confers superior assay performance under fast thermal cycling conditions, allowing more samples to be run in a day with the highest confidence, ideal for high‑throughput assays.