Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Red ITR Base 98.0+%, TCI America™
SDP

Catalog No. F008925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F008925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F008925G Supplier TCI America Supplier No. F008925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-35-8
Molecular Formula C11H18N2O3S
Molecular Weight (g/mol) 258.34
MDL Number MFCD00009045
InChI Key WBGVVXSCGNGJFL-UHFFFAOYSA-N
Synonym Fast Red ITR, Azoic Diazo Component 42, N1,N1-Diethyl-4-methoxymetanilamide, 3-Amino-N,N-diethyl-4-methoxybenzenesulfonamide
PubChem CID 66804
IUPAC Name 3-amino-N,N-diethyl-4-methoxybenzene-1-sulfonamide
SMILES CCN(CC)S(=O)(=O)C1=CC=C(OC)C(N)=C1

Specifications

Melting Point 105°C
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 258.34
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Fast Red ITR Base
EINECSNumber (3)-2038&(5)-2325
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.