Learn More
Fast Red TR Salt 1,5-Naphthalenedisulfonate Salt, Sigma-Aldrich
Catalog No. 01258841 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
01-258-841 5 g
1 options
Catalog No. 01-258-841 Supplier MilliporeSigma Supelco Supplier No. F67605G
Fast Red TR Salt 1,5-naphthalenedisulfonate salt has been used for tartrate-resistant acid phosphatase (TRAP) staining in tissue sections. It has been used as a chromogen in APAAP (alkaline phosphatase-anti-alkaline phosphatase) procedure.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 g
Product Type Stain Kit