Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Red Violet LB Salt, Sigma-Aldrich

Catalog No. 01259013 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-013 5 g
01-258-927 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-013 Supplier MilliporeSigma Supelco Supplier No. F33815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for quantitating alkaline phosphatase through histochemical, hematological, histological, and flow cytometric methods.

Fast Red Violet LB Salt, also known as 4'-amino-2'-chloro-5'-methylbenzanilide. It is a lipophilic amine with an aromatic system of moderate size and a hydrophilic diazonium cation. The diazonium salt is available commercially as a zinc chloride double salt. It comprises ≥90% dye content.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥90 %
Quantity 5 g
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.