Learn More
MSC IRWIN VISE-GRIP Locking Pliers, Curved Jaw, 125 mm, 1 ea
Catalog No. 5000509190
Encompass
These locking pliers feature a self-energizing lower jaw designed for maximum clamping force, ensuring three times more gripping power without slipping or stripping when working with various nut or bolt head styles. The 5 in. (125 mm) tool is engineered for durability and reliable performance in demanding applications.
  • Self-energizing lower jaw provides three times more gripping power
  • Designed to prevent slipping or stripping on various fasteners
  • Jaw capacity of 1-1/8 in. (29 mm)
  • Jaw thickness of 6.4 mm
Catalog No. 50-005-09190 Supplier MSC Supplier No. 98077191
These locking pliers feature a self-energizing lower jaw designed for maximum clamping force, ensuring three times more gripping power without slipping or stripping when working with various nut or bolt head styles. The 5 in. (125 mm) tool is engineered for durability and reliable performance in demanding applications.
  • Self-energizing lower jaw provides three times more gripping power
  • Designed to prevent slipping or stripping on various fasteners
  • Jaw capacity of 1-1/8 in. (29 mm)
  • Jaw thickness of 6.4 mm