Learn More
GOJO™ Fast Towels
SDP
Catalog No. 19270267 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Packaging Quantity:
4/Pk.
Catalog No. Packaging Quantity
19-270-267 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-270-267 Supplier GOJO™ Supplier No. 629804
Add to Cart
Add to Cart
This item is not returnable. View return policy
Multi-purpose wet towels for quick, convenient cleaning.
  • Large, textured wet shop towel.
  • All-in-one cleaning formula effective for cleaning hands, arms, tools and surfaces.
  • Contains skin moisturizers to help prevent dryness and irritation.
  • VOC-compliant formula.
  • For use anywhere heavy duty cleaning is required, no water needed.
  • Fresh citrus scent.
  • Effective on dirt, oil, and grease.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-purpose Towel
Length (Metric) 7.93 cm
Width (Metric) 7.93 cm
Container Type Bucket
Packaging Quantity 4/Pk.
Sterility Non-sterile