Learn More
INTACT GENOMICS INC FastAmp Plant Direct PCR Kit 1250 Reactions
Supplier:  INTACT GENOMICS INC 4615
Encompass
Intact Genomics FastAmp Plant Direct PCR kit is an easy and robust method to amplify DNA fragments from plant tissues with high specificity and high sensitivity without the need of complicated DNA purification steps. The advanced formulation of this kit also allows fast PCR cycling conditions without compromising PCR sensitivity, specificity and yield. The FastAmp PCR kit is an ideal and powerful tool for high-throughput genotyping, DNA amplification, and plant genome analysis.Highlights Direct PCR- no need to purify DNA Specially engineered Taq DNA polymerase with highest sensitivity and specificity Extremely short PCR protocol times Master mix format with premixed gel loading dye to reduce cross-contamination and sample handling errors Optimized for both low and high GC templatesApplications Genotyping Transgene detection Knockout analysis Sequencing
Catalog No. 50-000-83010