Learn More
INTACT GENOMICS INC FastAmp qRPA SYBR Kit, 25 Reactions
Catalog No. 5000082842
Encompass
IG FastAmp qRPA SYBR kit that contains ready-to-use master mixes with all components except primers and template for isothermal amplification, detection, and quantification of DNA using Recombinase Polymerase Amplification. With this kit, RPA progress can be monitored in real-time and quantified using standardized template concentrations. The FastAmp qRPA SYBR kit is compatible with high throughput (96-well or 384-well) format fluorescent readers for faster results in just 30 minutes at constant 37⁰C. Now, primer sets can be validated by screening in parallel for isothermal RPA assay optimization with Flexible endpoint detection compatibility. Kit Includes: ,FastAmp qRPA SYBR Reaction Master Mix, IG RPA Enzyme Mixture v2 (4x), 280 mM Magnesium Acetate (MgOAc), Nuclease-free Water. Storage Temperature: -20°C
Catalog No. 50-000-82842 Supplier INTACT GENOMICS INC Supplier No. 3611
IG FastAmp qRPA SYBR kit that contains ready-to-use master mixes with all components except primers and template for isothermal amplification, detection, and quantification of DNA using Recombinase Polymerase Amplification. With this kit, RPA progress can be monitored in real-time and quantified using standardized template concentrations. The FastAmp qRPA SYBR kit is compatible with high throughput (96-well or 384-well) format fluorescent readers for faster results in just 30 minutes at constant 37⁰C. Now, primer sets can be validated by screening in parallel for isothermal RPA assay optimization with Flexible endpoint detection compatibility. Kit Includes: ,FastAmp qRPA SYBR Reaction Master Mix, IG RPA Enzyme Mixture v2 (4x), 280 mM Magnesium Acetate (MgOAc), Nuclease-free Water. Storage Temperature: -20°C