Learn More
Bioworld FastBlot™ Transfer Buffer pH 11.0 (5X), 500 mL
Catalog No. 50255793
Encompass_Preferred
For electrotransfer of proteins to PVDF or nitrocellulose membranes. 1X Transfer Buffer contains 10 mM CHAPS pH 11 and 10% Methanol. Does not contain SDS.May be useful for large proteins or very basic proteins and for N-terminal sequencing applications. Commonly used with semi-dry transfer.pH (@ 25°C): 11
Catalog No. 50-255-793 Supplier Bioworld Supplier No. 403200621
For electrotransfer of proteins to PVDF or nitrocellulose membranes. 1X Transfer Buffer contains 10 mM CHAPS pH 11 and 10% Methanol. Does not contain SDS.May be useful for large proteins or very basic proteins and for N-terminal sequencing applications. Commonly used with semi-dry transfer.pH (@ 25°C): 11