Learn More
Bulldog Bio Inc FastGene Scriptase II ReadyMix (100 rxns), OdT & Random Hex-primers
SDP
Catalog No. NC1933460
Encompass
How would you construct the ideal reverse transcriptase? It would have to make very long cDNAs at high yields. It would have to be able to work from very small amounts of input RNA. And of course, it would have to work fast. FastGene Scriptase II checks all three boxes, and it does it at a reasonable price. Here’s how. First, mutations are inserted into the RNase H domain of native MuLV‘s reverse transcriptase delivering a higher yield of full-length cDNAs. Next, changes are made to increase the enzymes natural thermal stability providing a more robust enzyme that doesn’t lose processing power, even at elevated temperatures. Finally, the reaction buffer composition is optimized for high speed production of cDNAs from incredibly low levels of input RNA. This unique combination of size, speed, and production makes FastGene Scriptase II the optimal choice for even the most complex applications requiring cDNAs, such as RT-qPCR and NGS.
Catalog No. NC1933460 Supplier Bulldog Bio Inc Supplier No. LS65
How would you construct the ideal reverse transcriptase? It would have to make very long cDNAs at high yields. It would have to be able to work from very small amounts of input RNA. And of course, it would have to work fast. FastGene Scriptase II checks all three boxes, and it does it at a reasonable price. Here’s how. First, mutations are inserted into the RNase H domain of native MuLV‘s reverse transcriptase delivering a higher yield of full-length cDNAs. Next, changes are made to increase the enzymes natural thermal stability providing a more robust enzyme that doesn’t lose processing power, even at elevated temperatures. Finally, the reaction buffer composition is optimized for high speed production of cDNAs from incredibly low levels of input RNA. This unique combination of size, speed, and production makes FastGene Scriptase II the optimal choice for even the most complex applications requiring cDNAs, such as RT-qPCR and NGS.