Learn More
BioVendor, LLC FASTIN ELASTIN 110 ASSAYS STNDSZE
SDP
Catalog No. 501725946
Encompass
TypeColorimetric assayApplicationsCell culture and/or animal studies, Tissue extract, CellSample RequirementsA sample volume of between 50 and 500 μl is required, containing not less than 5 μg and not more than 70 μg elastin.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at RT or 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range5 – 70 μgLimit of Detection5 μg
Catalog No. 50-172-5946 Supplier BioVendor, LLC Supplier No. F2000
TypeColorimetric assayApplicationsCell culture and/or animal studies, Tissue extract, CellSample RequirementsA sample volume of between 50 and 500 μl is required, containing not less than 5 μg and not more than 70 μg elastin.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at RT or 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range5 – 70 μgLimit of Detection5 μg