Learn More
Aviva Systems Biology Corp FASTKD2 Antibody - middle region (ARP55108_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55108_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FASTKD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FASTKD2. Alternate names: KIAA0971ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-53945