Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastPrep-24, Second Year Extended Limited Warranty

Catalog No. MP116002599 Shop All MP Biomedicals, Inc Products
Catalog No. MP116002599 Supplier MP Biomedicals™ Supplier No. 116002599

missing translation for 'validMainframeMsg'

Warranty can be purchased anytime before the one year new instrument standard manufacturer's warranty expires.

Covers manufacturer's defects and includes repair or replacement of such defects, including transportation of instrument to be repaired or replaced without charges. Does not cover normal wear and tear or replacement of electrical cord, springs, rubber cups, dome, tubes holders, tube convers or other accessories.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) FastPrep 24 Classic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.