Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastRNA™ Win Kit

Catalog No. MP116080050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116080050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116080050 Supplier MP Biomedicals™ Supplier No. 116080050

missing translation for 'validMainframeMsg'

For easy and quick isolation of total RNA from a wide variety of plant samples

  • Designed to isolate and purify high quality, total RNA from plant cells and tissues for better RT-PCR results
  • The use of the FastPrep™ instrument combined with unique lysis buffers guarantees an efficient lysis of the starting material and a simultaneous inactivation of endogenous RNases
  • Genomic DNA is separated from the total RNA by binding to specially optimized mineral carrier particles included in the lysis buffer
  • RNA is then mixed with ethanol and transferred to the RNA binding membrane
  • Total RNA then binds, contaminants are washed away, and the pure RNA is eluted
  • No phenol or chloroform is required
  • Includes protocol for simultaneous isolation of proteins
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
For Use With (Application) Northern Blot, RNA Dot Blots, In-Vitro Translation, RT-PCR, ddRT-PCR, cDNA-Libraries, and TaqMan&Trade; Analysis and Array Technologies
No. of Reactions 50
Nucleic Acid Type RNA
Product Type FastRNA™ Win Kit
Quantity 50 Preps
Sample Source Plant Tissues
Sterility Non-sterile
WARNING: Cancer - www.P65Warnings.ca.gov

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.