Learn More
Cell Signaling Technology FastScan™ LbCpf1/Cas12a ELISA Kit, 480 Assays
SDP
Catalog No. 503134229
Encompass
The FastScan™ LbCpf1 (Strain ND2006)/Cas12a ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of LbCpf1 (Strain ND2006)/Cas12a protein. The assay involves incubating samples with a capture antibody and an HRP-linked detection antibody to form a sandwich complex immobilized on the plate. Following washes, TMB substrate is added, and the resulting signal is proportional to the protein quantity present.
  • Detects endogenous LbCpf1/Cas12a protein levels via sandwich ELISA
  • Validated for use across all species
  • Includes cell extraction buffer, TMB substrate, and wash buffer
  • Requires 4 washes at step 6 of the protocol
  • Utilizes microplate format for protein quantification
Catalog No. 50-313-4229 Supplier Cell Signaling Technology Supplier No. 88197V1
The FastScan™ LbCpf1 (Strain ND2006)/Cas12a ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of LbCpf1 (Strain ND2006)/Cas12a protein. The assay involves incubating samples with a capture antibody and an HRP-linked detection antibody to form a sandwich complex immobilized on the plate. Following washes, TMB substrate is added, and the resulting signal is proportional to the protein quantity present.
  • Detects endogenous LbCpf1/Cas12a protein levels via sandwich ELISA
  • Validated for use across all species
  • Includes cell extraction buffer, TMB substrate, and wash buffer
  • Requires 4 washes at step 6 of the protocol
  • Utilizes microplate format for protein quantification