Learn More
Cell Signaling Technology FastScan Phospho-Akt1 ELISA Kit, Ser473, Human/Mouse/Rat/Monkey, 5 Kit
SDP
Catalog No. 503132925
Encompass
The FastScan™ Phospho-Akt1 (Ser473) ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of Akt1 when phosphorylated at Ser473. The assay functions by incubating samples with a proprietary tagged capture antibody and an HRP-linked detection antibody, forming a sandwich with the target protein that is immobilized on the plate via an anti-tag antibody. The intensity of the signal measured at 450 nm is proportional to the amount of phospho-Akt1 (Ser473) present in the sample.
  • Validated for use with human, mouse, rat, and monkey samples.
  • Compatible with plate shakers for efficient incubation.
  • Includes a positive control for assay validation.
  • Custom antibody formulations ensure high specificity and sensitivity.
  • Supports rapid analysis with TMB substrate and colorimetric detection.
Catalog No. 50-313-2925 Supplier Cell Signaling Technology Supplier No. 54336V1
The FastScan™ Phospho-Akt1 (Ser473) ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of Akt1 when phosphorylated at Ser473. The assay functions by incubating samples with a proprietary tagged capture antibody and an HRP-linked detection antibody, forming a sandwich with the target protein that is immobilized on the plate via an anti-tag antibody. The intensity of the signal measured at 450 nm is proportional to the amount of phospho-Akt1 (Ser473) present in the sample.
  • Validated for use with human, mouse, rat, and monkey samples.
  • Compatible with plate shakers for efficient incubation.
  • Includes a positive control for assay validation.
  • Custom antibody formulations ensure high specificity and sensitivity.
  • Supports rapid analysis with TMB substrate and colorimetric detection.