Learn More
Cell Signaling Technology FastScan™ Phospho-SMAD2 ELISA Kit, Human, Mouse, Monkey, 5 Kit
SDP
Catalog No. 503134289
Encompass
The FastScan™ Phospho-SMAD2 (Ser465/467) ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of SMAD2 when phosphorylated at Ser465/467. The assay functions by incubating samples with a capture antibody and an HRP-linked detection antibody, forming a complex immobilized to the plate via an anti-tag antibody. The intensity of the signal generated after TMB substrate addition is proportional to the concentration of phospho-SMAD2 (Ser465/467) in the sample.
  • Provides high-sensitivity detection of phosphorylated SMAD2 (Ser465/467).
  • Utilizes a sandwich ELISA format for reliable and specific protein quantification.
  • Validated for use with human, mouse, and monkey samples.
  • Includes all necessary reagents for a complete assay workflow.
  • Enables custom plate configurations with included strip-well format.
Catalog No. 50-313-4289 Supplier Cell Signaling Technology Supplier No. 86932V1
The FastScan™ Phospho-SMAD2 (Ser465/467) ELISA Kit is a sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of SMAD2 when phosphorylated at Ser465/467. The assay functions by incubating samples with a capture antibody and an HRP-linked detection antibody, forming a complex immobilized to the plate via an anti-tag antibody. The intensity of the signal generated after TMB substrate addition is proportional to the concentration of phospho-SMAD2 (Ser465/467) in the sample.
  • Provides high-sensitivity detection of phosphorylated SMAD2 (Ser465/467).
  • Utilizes a sandwich ELISA format for reliable and specific protein quantification.
  • Validated for use with human, mouse, and monkey samples.
  • Includes all necessary reagents for a complete assay workflow.
  • Enables custom plate configurations with included strip-well format.