Learn More
Cell Signaling Technology PathScan™ RP Total Jak2 Sandwich ELISA Kit, Human, Mouse, 96 Assays
SDP
Catalog No. 503132511
Encompass
This sandwich ELISA kit is a solid-phase, rapid-protocol assay designed for the detection of endogenous levels of Total Jak2 protein in cell lysates. The assay achieves high-sensitivity results in a reduced timeframe of 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on a pre-coated microwell plate. The magnitude of the signal generated by the TMB substrate is directly proportional to the quantity of Total Jak2 present in the sample.
  • Solid-phase sandwich ELISA format for efficient protein quantification.
  • Rapid 1.5-hour assay time for accelerated workflow.
  • Custom antibody formulations specific to the kit requirements.
  • Suitable for detection in human and mouse species.
  • Includes strip-well plate design for flexible, custom configurations.
Catalog No. 50-313-2511 Supplier Cell Signaling Technology Supplier No. 56355V2
Add to Cart
Add to Cart
Request a Quote
This sandwich ELISA kit is a solid-phase, rapid-protocol assay designed for the detection of endogenous levels of Total Jak2 protein in cell lysates. The assay achieves high-sensitivity results in a reduced timeframe of 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on a pre-coated microwell plate. The magnitude of the signal generated by the TMB substrate is directly proportional to the quantity of Total Jak2 present in the sample.
  • Solid-phase sandwich ELISA format for efficient protein quantification.
  • Rapid 1.5-hour assay time for accelerated workflow.
  • Custom antibody formulations specific to the kit requirements.
  • Suitable for detection in human and mouse species.
  • Includes strip-well plate design for flexible, custom configurations.