Learn More
Cell Signaling Technology PathScan™ RP Total Jak2 Sandwich ELISA Kit, Human, Mouse, 480 Assays
SDP
Catalog No. 503131556
Encompass
The PathScan™ RP Total Jak2 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA) designed for the detection of endogenous levels of Jak2 protein in a reduced assay time of 1.5 hours. In this assay, cell lysates and detection antibody are incubated on the coated microwell plate in a single step to form a sandwich with the target protein. Following extensive washing, TMB reagent is added to generate a signal, with the magnitude of absorbance being proportional to the quantity of total Jak2 present in the sample.
  • Rapid 1.5 hour assay protocol
  • Specific detection of endogenous Jak2 protein
  • Validated for human and mouse species reactivity
  • Custom formulation antibodies optimized for kit performance
  • Includes 12 x 8-well strip plate for custom configurations
Catalog No. 50-313-1556 Supplier Cell Signaling Technology Supplier No. 56355V1
The PathScan™ RP Total Jak2 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA) designed for the detection of endogenous levels of Jak2 protein in a reduced assay time of 1.5 hours. In this assay, cell lysates and detection antibody are incubated on the coated microwell plate in a single step to form a sandwich with the target protein. Following extensive washing, TMB reagent is added to generate a signal, with the magnitude of absorbance being proportional to the quantity of total Jak2 present in the sample.
  • Rapid 1.5 hour assay protocol
  • Specific detection of endogenous Jak2 protein
  • Validated for human and mouse species reactivity
  • Custom formulation antibodies optimized for kit performance
  • Includes 12 x 8-well strip plate for custom configurations