Learn More
Cell Signaling Technology FastScan ELISA Kit, Total p44/42 MAPK, 480 Assays
SDP
Catalog No. 503134124
Encompass
The FastScan™ Total p44/42 MAPK (Erk1/2) ELISA Kit is a sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of p44/42 MAPK (Erk1/2) in human, mouse, rat, and monkey samples. The assay utilizes a capture antibody conjugated with a proprietary tag and an HRP-linked detection antibody, forming a sandwich complex immobilized to the plate via an anti-tag antibody. The resulting signal magnitude is proportional to the target protein quantity.
  • Validated for use with human, mouse, rat, and monkey samples
  • Features a streamlined sandwich ELISA protocol
  • Includes optimized capture and detection antibodies in custom formulations
  • Designed for high-sensitivity detection of total p44/42 MAPK
  • Compatible with plate-based colorimetric detection at 450 nm
Catalog No. 50-313-4124 Supplier Cell Signaling Technology Supplier No. 67404V1
The FastScan™ Total p44/42 MAPK (Erk1/2) ELISA Kit is a sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of p44/42 MAPK (Erk1/2) in human, mouse, rat, and monkey samples. The assay utilizes a capture antibody conjugated with a proprietary tag and an HRP-linked detection antibody, forming a sandwich complex immobilized to the plate via an anti-tag antibody. The resulting signal magnitude is proportional to the target protein quantity.
  • Validated for use with human, mouse, rat, and monkey samples
  • Features a streamlined sandwich ELISA protocol
  • Includes optimized capture and detection antibodies in custom formulations
  • Designed for high-sensitivity detection of total p44/42 MAPK
  • Compatible with plate-based colorimetric detection at 450 nm