Learn More
Cell Signaling Technology FastScan™ Tri-Methyl-Histone H3 (Lys27) ELISA Kit, Human/Mouse/Rat/Monkey, 20 Kit
SDP
Catalog No. 503132276
Encompass
This sandwich enzyme-linked immunosorbent assay (ELISA) kit is designed for the detection of endogenous levels of histone H3 when tri-methylated at Lys27. The assay functions by incubating samples with a capture antibody conjugated with a proprietary tag and a detection antibody linked to HRP, forming a sandwich with the target protein in solution. This complex is immobilized to the plate via an anti-tag antibody, and the resulting signal intensity is directly proportional to the quantity of tri-methyl-histone H3 (Lys27) present.
  • Detects endogenous levels of tri-methyl-histone H3 (Lys27)
  • Validated for reactivity in human, mouse, rat, and monkey samples
  • Enables sensitive protein quantification
  • Requires 4 washes at step 6 of the protocol
  • Store at 4°C
Catalog No. 50-313-2276 Supplier Cell Signaling Technology Supplier No. 15177V3
Add to Cart
Add to Cart
Request a Quote
This sandwich enzyme-linked immunosorbent assay (ELISA) kit is designed for the detection of endogenous levels of histone H3 when tri-methylated at Lys27. The assay functions by incubating samples with a capture antibody conjugated with a proprietary tag and a detection antibody linked to HRP, forming a sandwich with the target protein in solution. This complex is immobilized to the plate via an anti-tag antibody, and the resulting signal intensity is directly proportional to the quantity of tri-methyl-histone H3 (Lys27) present.
  • Detects endogenous levels of tri-methyl-histone H3 (Lys27)
  • Validated for reactivity in human, mouse, rat, and monkey samples
  • Enables sensitive protein quantification
  • Requires 4 washes at step 6 of the protocol
  • Store at 4°C