Learn More
Sigma Aldrich Fine Chemicals Biosciences FastStartTM PCR Master
Catalog No. 502907396
Encompass
Hot start polymerase chain reaction (PCR) is effortless using the FastStart(TM) PCR Master a ready-to-use double-concentrated hot start master mix. This master mix contains FastStart Taq DNA Polymerase Roche high quality PCR-Grade nucleotides and all other reagents required for PCR and two-step reverse transcriptase (RT)-PCR on thermal cycler instruments.FastStart(TM) PCR Master accepts modified nucleotides such as Digoxigenin (DIG)-dUTP. Labeling with DIG-dUTP can be achieved by adding the modified nucleotide in a final concentration of 10 muM to the final volume.Combine this master mix with Roches Transcriptor First Strand cDNA Synthesis Kit to achieve excellent results in two-step RT-PCR.
Catalog No. 50-290-7396 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4710444001
Hot start polymerase chain reaction (PCR) is effortless using the FastStart(TM) PCR Master a ready-to-use double-concentrated hot start master mix. This master mix contains FastStart Taq DNA Polymerase Roche high quality PCR-Grade nucleotides and all other reagents required for PCR and two-step reverse transcriptase (RT)-PCR on thermal cycler instruments.FastStart(TM) PCR Master accepts modified nucleotides such as Digoxigenin (DIG)-dUTP. Labeling with DIG-dUTP can be achieved by adding the modified nucleotide in a final concentration of 10 muM to the final volume.Combine this master mix with Roches Transcriptor First Strand cDNA Synthesis Kit to achieve excellent results in two-step RT-PCR.