Learn More
Sigma Aldrich Fine Chemicals Biosciences fastStartTM Taq DNA Polyme
Catalog No. 502914460
Encompass
The enzyme dNTPack comprises fastStart Taq DNA Polymerase and a ready-to-use SOLUTIONon of PCR grade nucleotides. fastStart Taq DNA Polymerase is a versatile enzyme that can be used in a wide variety of applications and on multiple instrument platforms. This modified thermostable recombinant Taq DNA polymerase is inactive at temperatures below 75 C but is activated by a 2 to 4 minute heat activation step at 95 C. Since it is inactive at low temperatures fastStart Taq DNA Polymerase cannot elongate non-specific primer-template hybrids that may form at those temperatures. An optimized PCR buffer system and a GC-RICH SOLUTIONon to enable the enzyme to handle a wide range of templates are also supplied.
Catalog No. 50-291-4460 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4738381001
The enzyme dNTPack comprises fastStart Taq DNA Polymerase and a ready-to-use SOLUTIONon of PCR grade nucleotides. fastStart Taq DNA Polymerase is a versatile enzyme that can be used in a wide variety of applications and on multiple instrument platforms. This modified thermostable recombinant Taq DNA polymerase is inactive at temperatures below 75 C but is activated by a 2 to 4 minute heat activation step at 95 C. Since it is inactive at low temperatures fastStart Taq DNA Polymerase cannot elongate non-specific primer-template hybrids that may form at those temperatures. An optimized PCR buffer system and a GC-RICH SOLUTIONon to enable the enzyme to handle a wide range of templates are also supplied.