Learn More
Sigma Aldrich Fine Chemicals Biosciences fastStartTM Taq DNA Polyme
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 12032902001
Encompass
fastStart(TM) Taq DNA Polymerase is a versatile enzyme that can be used in a wide variety of applications and on multiple instrument platforms. This modified thermostable recombinant Taq DNA polymerase is inactive at temperatures below 75 C but is activated by a 2 to 4 minute heat activation step at 95 C. Since it is inactive at low temperatures fastStart(TM) Taq DNA Polymerase cannot elongate non-specific primer-template hybrids that may form at those temperatures. fastStart(TM) Taq DNA Polymerase is an ideal tool for hot start PCR because the enzyme remains inactive during PCR set-up and prior to the initial denaturation step.
Catalog No. 50-290-0140