Learn More
Electron Microscopy Sciences FastWells™ Fastwell 1 Incubation Chamber, 32x19 mmx1.0 mm
SDP
Catalog No. 5026560
Encompass_Preferred
FastWells stickable, stackable, sealable, peelable and reusealble reagent barriers. Sticky, flexible silicone gaskets form hydrophobic reagent barriers around specimens without messy adhesives or special slides. May be stacked to increase depth and volume. They remain sealed during agitated washing steps and can be washed, autoclaved and reused. Can also be sealed to form incubation chambers using flexible HybriSlip™ Covers. 32mm x 19mm x 1.0mm Depth, 25.5mm x 44mm OD
Catalog No. 50-265-60 Supplier Electron Microscopy Sciences Supplier No. 7032554
FastWells stickable, stackable, sealable, peelable and reusealble reagent barriers. Sticky, flexible silicone gaskets form hydrophobic reagent barriers around specimens without messy adhesives or special slides. May be stacked to increase depth and volume. They remain sealed during agitated washing steps and can be washed, autoclaved and reused. Can also be sealed to form incubation chambers using flexible HybriSlip™ Covers. 32mm x 19mm x 1.0mm Depth, 25.5mm x 44mm OD