Learn More
Electron Microscopy Sciences FastWells™ 1 Incubation Chamber, 20 mm dia x1.0 mm deep
SDP
Catalog No. 5026561
Encompass_Preferred
FastWells stickable, stackable, sealable, peelable and reusealble reagent barriers. Sticky, flexible silicone gaskets form hydrophobic reagent barriers around specimens without messy adhesives or special slides. May be stacked to increase depth and volume. They remain sealed during agitated washing steps and can be washed, autoclaved and reused. Can also be sealed to form incubation chambers using flexible HybriSlip™ Covers. 20mm Dia x 1.0mm Depth, 25mm x 25mm OD
Catalog No. 50-265-61 Supplier Electron Microscopy Sciences Supplier No. 7032552
FastWells stickable, stackable, sealable, peelable and reusealble reagent barriers. Sticky, flexible silicone gaskets form hydrophobic reagent barriers around specimens without messy adhesives or special slides. May be stacked to increase depth and volume. They remain sealed during agitated washing steps and can be washed, autoclaved and reused. Can also be sealed to form incubation chambers using flexible HybriSlip™ Covers. 20mm Dia x 1.0mm Depth, 25mm x 25mm OD