Learn More
Research Products International Corp Fastwells, 20 mm Diameter, 50ul Volume, 50 per Package
SDP
Catalog No. 50212664
Encompass
Fastwells form hydrophobic reagent barriers around various size specimens. Ideal for rapid isolation of cells and tissues on slides or coverslips for antibody incubations. Fastwells remain sealed during agitated washing steps to prevent leakage. High tacky silicone material peels off quickly without leaving a residue. Fastwells can be washed and autoclaved.
Catalog No. 50-212-664 Supplier Research Products International Corp Supplier No. 248936
Add to Cart
Add to Cart
Fastwells form hydrophobic reagent barriers around various size specimens. Ideal for rapid isolation of cells and tissues on slides or coverslips for antibody incubations. Fastwells remain sealed during agitated washing steps to prevent leakage. High tacky silicone material peels off quickly without leaving a residue. Fastwells can be washed and autoclaved.