Learn More
FASTER CHEMISTRY LLC Faster Chemistry FastWoRX-M 100 g
Supplier:  FASTER CHEMISTRY LLC FASTWORXM100
Encompass
FastWoRX Absorbent Powder - 100 g per bottle. FastWoRX products are hydrophobic powders that absorb most organic compounds. With them, liquid-liquid extraction (LLE) for reaction work-ups can be replaced by a simple filtration or magnetic separation. Work-ups can be done in half the time or less. Solvent use can be reduced by 90% or more. Emulsions cannot form. Automation of FastWoRX-based work-ups is much easier than automating LLE.
Catalog No. NC1569195